Organophosphorus compounds
Phosphate triéthylique, 98 %, Thermo Scientific Chemicals
CAS: 122-52-1 Poids moléculaire (g/mol): 166.16 Numéro MDL: MFCD00009084 Clé InChI: BDZBKCUKTQZUTL-UHFFFAOYSA-N Synonyme: c2h5o 3p, p oet 3, hsdb 895, unii-6b2r04s55g, fosforyn trojetylowy czech, phosphorous acid triethyl ester, triethylphosphite, ethyl phosphite, eto 3p, triethoxyphosphine, phosphorous acid, triethyl ester CID PubChem: 31215 Nom IUPAC: phosphite de triéthyle SMILES: CCOP(OCC)OCC
Phosphite de triéthyle, 98 %, Thermo Scientific Chemicals
CAS: 122-52-1 Formule moléculaire: C6H15O3P Poids moléculaire (g/mol): 166.157 Numéro MDL: MFCD00009084 Clé InChI: BDZBKCUKTQZUTL-UHFFFAOYSA-N Synonyme: c2h5o 3p, p oet 3, hsdb 895, unii-6b2r04s55g, fosforyn trojetylowy czech, phosphorous acid triethyl ester, triethylphosphite, ethyl phosphite, eto 3p, triethoxyphosphine, phosphorous acid, triethyl ester CID PubChem: 31215 Nom IUPAC: phosphite de triéthyle SMILES: CCOP(OCC)OCC
Isopropyldiphénylphosphine, 99 %, Thermo Scientific Chemicals
CAS: 6372-40-3 Formule moléculaire: C15H17P Poids moléculaire (g/mol): 228.28 Numéro MDL: MFCD00015028 Clé InChI: LLZAIAIZAVMQIG-UHFFFAOYSA-N Synonyme: i-propyldiphenylphosphine, diphenyl propan-2-yl phosphane, isopropyl-diphenylphosphine, acmc-209nho, isopropyl diphenyl phosphine, isopropyldiphenylphosphane, phosphine, 1-methylethyl diphenyl, isopropyldiphenylphosphine CID PubChem: 80754 Nom IUPAC: diphényle(propane-2-yl)phosphane SMILES: CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1
Méthyldiphénylphosphine, 99 %, Thermo Scientific Chemicals
CAS: 1486-28-8 Formule moléculaire: C13H13P Poids moléculaire (g/mol): 200.22 Numéro MDL: MFCD00008508 Clé InChI: UJNZOIKQAUQOCN-UHFFFAOYSA-N Synonyme: methyldi-phenylphosphine, methyl diphenylphosphine, pubchem6421, methyldiphenylphoshine, diphenylphosphinomethane, methyldiphenylphsphine, phosphine, methyldiphenyl, methyl diphenyl phosphine, diphenylmethylphosphine, methyldiphenylphosphine CID PubChem: 73879 Nom IUPAC: méthyl(diphényl)phosphane SMILES: CP(C1=CC=CC=C1)C1=CC=CC=C1
Phosphite triisopropylique, 96 %, Thermo Scientific Chemicals
CAS: 116-17-6 Formule moléculaire: C9H21O3P Poids moléculaire (g/mol): 208.24 Clé InChI: SJHCUXCOGGKFAI-UHFFFAOYSA-N Synonyme: tri-i-propylphosphite, triisopropylphosphite, unii-rj7qu4yqun, tri-2-propylphosphite, phosphorous acid, triisopropyl ester, triisopropoxyphosphine, isopropyl phosphite, tri, phosphorous acid, tris 1-methylethyl ester, tri-2-propyl phosphite, triisopropyl phosphite CID PubChem: 8304 Nom IUPAC: Phosphite de tripropan-2-yle SMILES: CC(C)OP(OC(C)C)OC(C)C
Tert-butyldichlorophosphine, 98 %, Thermo Scientific Chemicals
CAS: 25979-07-1 Formule moléculaire: C4H9Cl2P Poids moléculaire (g/mol): 158.99 Numéro MDL: MFCD00013615 Clé InChI: NMJASRUOIRRDSX-UHFFFAOYSA-N Synonyme: tert-butylphosphine dichloride, tert-butyl dichloro phosphine, acmc-1cmec, pubchem6471, snpladsptpdwuup@, phosphonous dichloride, 1,1-dimethylethyl, tert-butyl dichloro phosphane, tert-c4h9pcl2, t-butyldichlorophosphine, tert-butyldichlorophosphine CID PubChem: 117690 Nom IUPAC: tert-butyl(dichloro)phosphane SMILES: CC(C)(C)P(Cl)Cl
Chloro(diéthyl)phosphine, Thermo Scientific Chemicals
CAS: 686-69-1 Formule moléculaire: C4H10ClP Poids moléculaire (g/mol): 124.548 Numéro MDL: MFCD00013621 Clé InChI: INJBDKCHQWVDGT-UHFFFAOYSA-N Synonyme: chlorodiethyl phosphine, diethylphosphinous chloride #, diethylphosphinous chloride, acmc-1avkn, chloro diethyl phosphane, pubchem6480, phosphinous chloride, diethyl, chloro diethyl phosphine, diethylchlorophosphine, chlorodiethylphosphine CID PubChem: 69639 Nom IUPAC: chloro(diéthyl)phosphane SMILES: CCP(CC)Cl
Dichloro[bis(1,3-diphénylphosphino)propane]nickel(II), Thermo Scientific Chemicals
CAS: 15629-92-2 Formule moléculaire: C27H26Cl2NiP2 Poids moléculaire (g/mol): 542.04 Numéro MDL: MFCD00015318 Clé InChI: REQVOKPBMSCTNI-UHFFFAOYSA-L Synonyme: 1,3-Bis(diphenylphosphino)propane nickel(II) chloride CID PubChem: 131675641 Nom IUPAC: 3-diphénylphosphanylpropyl(diphényl)phosphane ; nickel ; dichlorhydrate SMILES: Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Dicyclohexylphénylphosphine, 95 %, Thermo Scientific Chemicals
CAS: 6476-37-5 Formule moléculaire: C18H27P Poids moléculaire (g/mol): 274.39 Numéro MDL: MFCD00003854 Clé InChI: VPLLTGLLUHLIHA-UHFFFAOYSA-N Synonyme: dicyclohexyl-phenyl-phosphane, acmc-1bey7, pphcy2, dicyclohexyl phenyl phosphane, phenylphosphinodicyclohexane, phenyldicyclohexylphosphine, phosphine, dicyclohexylphenyl, dicyclohexyl phenyl phosphine, dicyclohexylphenylphosphine CID PubChem: 80970 Nom IUPAC: dicyclohexyl(phényl)phosphane SMILES: C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1
Tribenzylphosphine, Thermo Scientific Chemicals
CAS: 7650-89-7 Formule moléculaire: C21H21P Poids moléculaire (g/mol): 304.37 Numéro MDL: MFCD00014439 Clé InChI: IFXORIIYQORRMJ-UHFFFAOYSA-N Synonyme: tribenzylphosphine 1g, acmc-20ajfj, phosphine, tribenzyl, tris phenylmethyl phosphine, tris phenylmethyl phosphane, tribenzyl phosphine, phosphine, tris phenylmethyl, tribenzylphosphine CID PubChem: 82101 Nom IUPAC: tribenzylphosphane SMILES: C(P(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Bis[2-(di-tert-butylphosphino)éthyl]amine, solution 10 % p/w dans du THF, Thermo Scientific Chemicals
CAS: 944710-34-3 Formule moléculaire: C20H45NP2 Poids moléculaire (g/mol): 361.54 Numéro MDL: MFCD17014018 Clé InChI: MJYKYLNPIGDVEF-UHFFFAOYSA-N Synonyme: bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-t-butylphosphino ethyl amine CID PubChem: 51346231 Nom IUPAC: 2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanyléthyl)éthanamine SMILES: CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C
Chloro(diéthyl)phosphine, 95 %, Thermo Scientific Chemicals
CAS: 686-69-1 Formule moléculaire: C4H10ClP Poids moléculaire (g/mol): 124.55 Numéro MDL: MFCD00013621 Clé InChI: INJBDKCHQWVDGT-UHFFFAOYSA-N Synonyme: chlorodiethyl phosphine, diethylphosphinous chloride #, diethylphosphinous chloride, acmc-1avkn, chloro diethyl phosphane, pubchem6480, phosphinous chloride, diethyl, chloro diethyl phosphine, diethylchlorophosphine, chlorodiethylphosphine CID PubChem: 69639 Nom IUPAC: chloro(diéthyl)phosphane SMILES: CCP(CC)Cl
(2R,3R)-(+)-Bis(diphénylphosphino)butane, 98 %, Thermo Scientific Chemicals
CAS: 74839-84-2 Formule moléculaire: C28H28P2 Poids moléculaire (g/mol): 426.48 Numéro MDL: MFCD00075403 Clé InChI: FWXAUDSWDBGCMN-ZBXWWWQKNA-N Synonyme: r,?r ?-?chiraphos, 2r,3r-bis diphenylphosphino butane, phosphine, 1r,2r-1,2-dimethyl-1,2-ethanediyl bis diphenyl, cira, 2r,3r-+-2,3-bis diphenylphosphino butane, 2r,3r-butane-2,3-diylbis diphenylphosphine, 2r,3r-+-bis diphenylphosphino butane, unii-1t086b9q1j, chiraphos, r,r, r,r-chiraphos CID PubChem: 2724948 Nom IUPAC: [(2R,3R)-3-diphénylphosphanylbutan-2-yl]-diphénylphosphane SMILES: C[C@H]([C@@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Dicyclohexylphénylphosphine, 98 %, Thermo Scientific Chemicals
CAS: 6476-37-5 Formule moléculaire: C18H27P Poids moléculaire (g/mol): 274.39 Numéro MDL: MFCD00003854 Clé InChI: VPLLTGLLUHLIHA-UHFFFAOYSA-N Synonyme: dicyclohexyl-phenyl-phosphane, acmc-1bey7, pphcy2, dicyclohexyl phenyl phosphane, phenylphosphinodicyclohexane, phenyldicyclohexylphosphine, phosphine, dicyclohexylphenyl, dicyclohexyl phenyl phosphine, dicyclohexylphenylphosphine CID PubChem: 80970 Nom IUPAC: dicyclohexyl(phényl)phosphane SMILES: C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1
1,3-bis(diphénylphosphino)propane, 97 %, Thermo Scientific Chemicals
CAS: 6737-42-4 Formule moléculaire: C27H26P2 Poids moléculaire (g/mol): 412.45 Numéro MDL: MFCD00003050 Clé InChI: LVEYOSJUKRVCCF-UHFFFAOYSA-N Synonyme: 1,3-bis-diphenylphosphino propane, bis 1,3-diphenylphosphino propane, 1,3-bis diphenylphosphino propane dppp, 1,3-bis diphenyphosphino propane, 3-diphenylphosphanylpropyl diphenyl phosphane, 3-diphenylphosphanyl propyl diphenylphosphane, phosphine, 1,3-propanediylbis diphenyl, trimethylenebis diphenylphosphine, dppp, 1,3-bis diphenylphosphino propane CID PubChem: 81219 SMILES: C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Dichlorométhylphosphine, 90 %, Tech., Thermo Scientific Chemicals
CAS: 676-83-5 Formule moléculaire: CH3Cl2P Poids moléculaire (g/mol): 116.909 Numéro MDL: MFCD00013616 Clé InChI: CDPKWOKGVUHZFR-UHFFFAOYSA-N Synonyme: phosphine, dichloromethyl, methylphosphorus dichloride, methyl phosphonous dichloride, phosphonous dichloride, methyl, ch3pcl2, phosphonous dichloride, p-methyl, dichloro methyl phosphane, methylphosphonous dichloride, dichloromethylphosphine, methyldichlorophosphine CID PubChem: 61194 Nom IUPAC: dichloro(méthyl)phosphane SMILES: CP(Cl)Cl
Tri-n-butylphosphine, 95 %, Thermo Scientific Chemicals
CAS: 998-40-3 Formule moléculaire: C12H27P Poids moléculaire (g/mol): 202.32 Numéro MDL: MFCD00009462 Clé InChI: TUQOTMZNTHZOKS-UHFFFAOYSA-N Synonyme: tributyl phosphine tbp, tri-n-butyl phosphine, unii-0o52fjr7wn, tributyl-phosphane, tributylfosfin czech, tributylfosfin, tris butyl phosphine, phosphine, tributyl, tri-n-butylphosphine, tributylphosphine CID PubChem: 13831 Nom IUPAC: tributylphosphane SMILES: CCCCP(CCCC)CCCC
Chlorure de bis(triéthylphosphine)palladium(II), Thermo Scientific Chemicals
CAS: 28425-04-9 Formule moléculaire: C12H30Cl2P2Pd Poids moléculaire (g/mol): 413.64 Numéro MDL: MFCD00191831 Clé InChI: ULYNIEUXPCUIEL-UHFFFAOYSA-L Synonyme: bis triethylphosphine dichloridopalladium ii, dichloropalladium-triethylphosphane 1/2, palladium,dichlorobis triethylphosphine, pdcl2 pet3 2, bis triethylphosphine palladium ii chloride, trans-dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium dichloride, dichlorobis triethylphosphine palladium ii CID PubChem: 11144124 Nom IUPAC: dichloropalladium ; triéthylphosphane SMILES: Cl[Pd]Cl.CCP(CC)CC.CCP(CC)CC
Tris(2-cyanoéthyl)phosphine, 95 %, Thermo Scientific Chemicals
CAS: 4023-53-4 Formule moléculaire: C9H12N3P Poids moléculaire (g/mol): 193.19 Clé InChI: CHZAMJVESILJGH-UHFFFAOYSA-N Synonyme: tris 2-cyanoethyl phosphine 10g, propanenitrile,3',3-phosphinidynetris, 3-bis 2-cyanoethyl phosphanyl propanenitrile, p ch2ch2cn 3, acmc-20ajc3, propanenitrile, 3,3',3-phosphinidynetris, tris-2-cyanoethyl-phosphine, tris-2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris 2-cyanoethyl phosphine CID PubChem: 77639 Nom IUPAC: 3-[bis(2-cyanoéthyl)phosphanyl]propanenitrile SMILES: C(CP(CCC#N)CCC#N)C#N
Triisopropylphosphine, 90+ %, Thermo Scientific Chemicals
CAS: 6476-36-4 Formule moléculaire: C9H21P Poids moléculaire (g/mol): 160.241 Numéro MDL: MFCD00015029 Clé InChI: IGNTWNVBGLNYDV-UHFFFAOYSA-N Synonyme: pubchem6458, acmc-20ajdu, triisopropyl phosphine, trisisopropylphosphine, triisopropylphosphane, tri-isopropylphosphine, phosphine, tris 1-methylethyl, phosphine, triisopropyl, tri-i-propylphosphine, triisopropylphosphine CID PubChem: 80969 Nom IUPAC: Tri(propan-2-yl)phosphane SMILES: CC(C)P(C(C)C)C(C)C
Bis(diphénylphosphino)méthane, 97 %, Thermo Scientific Chemicals
CAS: 2071-20-7 Formule moléculaire: C25H22P2 Poids moléculaire (g/mol): 384.40 Numéro MDL: MFCD00003537 Clé InChI: XGCDBGRZEKYHNV-UHFFFAOYSA-N Synonyme: 1,1-bis diphenylphosphino methane, diphenylphosphanyl methyl diphenylphosphane, methanediylbis diphenylphosphine, unii-i5o2hzr38v, tetraphenylmethylenediphosphine, phosphine, methylenebis diphenyl, methylenebis diphenylphosphine, dppm, bis diphenylphosphino methane CID PubChem: 74952 Nom IUPAC: diphénylphosphanylméthyl(diphényl)phosphane SMILES: C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Chlorodiisopropylphosphine, 96 %, Thermo Scientific Chemicals
CAS: 40244-90-4 Formule moléculaire: C6H14ClP Poids moléculaire (g/mol): 152.61 Numéro MDL: MFCD00015027 Clé InChI: JZPDBTOWHLZQFC-UHFFFAOYSA-N Synonyme: diisopropylphosphinyl chloride, acmc-209jc6, chloro diisopropylphosphine, chlorodiisopropylphosphane, pubchem6476, ipr2pcl, phosphinous chloride, bis 1-methylethyl, di-i-propylchlorophosphine, diisopropylchlorophosphine, chlorodiisopropylphosphine CID PubChem: 538967 Nom IUPAC: chloro-di(propan-2-yl)phosphane SMILES: CC(C)P(C(C)C)Cl
Tris(2-cyanoéthyl)phosphine, 94 %, Thermo Scientific Chemicals
CAS: 4023-53-4 Formule moléculaire: C9H12N3P Poids moléculaire (g/mol): 193.19 Numéro MDL: MFCD00013828 Clé InChI: CHZAMJVESILJGH-UHFFFAOYSA-N Synonyme: tris 2-cyanoethyl phosphine 10g, propanenitrile,3',3-phosphinidynetris, 3-bis 2-cyanoethyl phosphanyl propanenitrile, p ch2ch2cn 3, acmc-20ajc3, propanenitrile, 3,3',3-phosphinidynetris, tris-2-cyanoethyl-phosphine, tris-2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris 2-cyanoethyl phosphine CID PubChem: 77639 Nom IUPAC: 3-[bis(2-cyanoéthyl)phosphanyl]propanenitrile SMILES: C(CP(CCC#N)CCC#N)C#N
Bis(di-tert-butyl-phénylphosphine)palladium(0), Pd 19,3 %, Thermo Scientific Chemicals
CAS: 52359-17-8 Formule moléculaire: C28H46P2Pd Poids moléculaire (g/mol): 551.04 Numéro MDL: MFCD15071400 Clé InChI: KJTQUBRAVOMHKY-UHFFFAOYSA-N Synonyme: bis di-tert-butyl phenyl phosphane palladium, di-tert-butyl phenyl phosphane-palladium 2/1, palladium, bis bis 1,1-dimethylethyl phenylphosphine CID PubChem: 71365671 Nom IUPAC: ditert-butyl(phényl)phosphane ; palladium SMILES: [Pd].CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
Bis(tricyclohexylphosphine)palladium(0), blanc cassé cristallin, Thermo Scientific Chemicals
CAS: 33309-88-5 Formule moléculaire: C36H66P2Pd Poids moléculaire (g/mol): 667.29 Numéro MDL: MFCD01073796 Clé InChI: JGBZTJWQMWZVNX-UHFFFAOYSA-N Synonyme: bis tricyclohexylphosphine-palladium o, palladium-tricyclohexylphosphine 1:2, palladium; tricyclohexylphosphane, pd pcy3 2, palladium, bis tricyclohexylphosphine, bis tricyclohexylphosphine palladium, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium 0 CID PubChem: 2734559 Nom IUPAC: palladium ; tricyclohexylphosphane SMILES: [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1