Agitateurs et barreaux d’agitation
Fisherbrand™ Stir Bar Box Set, Cylindrical
Provides general purpose stirring for a wide variety of applications. Fisherbrand™ Stir Bar Box Sets, Cylindrical are isostatically molded in pure, FDA-approved PTFE plastic.
Thermo Scientific™ RT Basic Series Magnetic Stirrers
Experience the reliability of our plug-and-play magnetic stirrers, ideal for routine stirring applications.
Fisherbrand™ PTFE Stir Bars with Pivot Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
Fisherbrand™ Plain Economy PTFE Stir Bar
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
Fisherbrand™ Stir Bar Box Set, Octahedral
For use in containers with bases that are slightly curved or uneven. Fisherbrand™ Star Bar Box Set, Octahedral is isostatically molded in pure, FDA-approved PTFE plastic.
Cole-Parmer™ Stuart™ UC152D Model Hot Plate Stirrer
This Stuart™ UC152D Model Hot Plate Stirrer features a glass ceramic top with excellent chemical resistance.
Fisherbrand™ Isotemp™ RT Advanced Hot Plate Stirrer, 350°C, Ceramic, Aluminum
Stirring Range: 30 to 2000 rpm; Shape: Round; Size: 5.5 in. dia.; Max. Sample Load: 25 kg; Plug Type: Various Styles
Thermo Scientific™ Cimarec i Poly 15 and Multipoint Stirrers
Conduct stirring applications with these stirrers, which are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Fisherbrand™ Color-coded Octahedral PTFE Stir Bar
Use when quick visual identification is highly important. Fisherbrand™ Color-coded Octahedral PTFE Stir Bar is available in three colors: blue, red, and yellow.
Azlon™ Pivot Ring Magnetic Stirrer Bar
Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.
Fisherbrand™ Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Bel-Art™ SP Scienceware™ Egg-Shaped Spinbar™ Magnetic Stir Bars
Designed for use in round-bottom vessels
Thermo Scientific™ Cimarec™ Mobil Direct Stirrers
Perform high-volume applications with the exceptional stirring power of these mobil direct stirrers.
IKA™ C-MAG HS 10 Magnetic Stirrer with Heating
Magnetic stirrer with heating and ceramic heating plate, which offers excellent chemical resistance.
Fisherbrand™ Isotemp™ RT Hot Plate Stirrer, 350°C, Aluminum, Ceramic
Stirring Range: 50 to 2000 rpm; Shape: Round; Size: 5.5 in. dia.; Max. Sample Load: 25 kg; Plug Type: Various Styles
Azlon™ Magnetic Retrievers for Stirrer Bars
Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. Azlon™ Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.
Fisherbrand™ Stir Bar Box Set, Oval
Guaranteed inert and leak-free. Fisherbrand™ Stir Bar Box Set, Oval is isostatically molded in pure, FDA-approved PTFE plastic.
Thermo Scientific™ SuperNuova+™ Stirring Hotplates Series
Get reliable performance, dependable safety and simple operation.
Thermo Scientific™ RT Touch Series Magnetic Stirrers
Stay in control with our low-profile, digital display stirrers, ideal for routine stirring applications where visible stir speed is required.
Thermo Scientific™ Cimarec™ Power Direct High Capacity Stirrer
Stir up to 40L with the integrated controller of the Thermo Scientific™ Cimarec™ Power Direct, ideal for high-capacity stirring.
Thermo Scientific™ RT2 Advanced Hotplate Stirrer
Experience precision with our low-profile, digital display hotplate stirrer, equipped with PT100 sensor for precise temperature measurements for sensitive experimental procedures.
Fisherbrand™ Oval PTFE Stir Bar
Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.
Fisherbrand™ Cylindrical PTFE or Glass Stir Bars
Alnico V magnet core encapsulated by PTFE or glass . Fisherbrand™ Cylindrical PTFE or Glass Stir Bars are autoclavable.
Fisherbrand™ Cylindrical PTFE Stir Bars
For use with small containers
BRAND™ PTFE Magnetic Stirring Bars with Pivot Ring, Cylindrical Shape
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars with Pivot Ring, Cylindrical Shape are for use with vessels that have slightly uneven bottoms. Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Cole-Parmer™ SHP-200-MP Stuart Three Position Stirring Hot Plate
SHP-200-MP Stuart Three Position Stirring Hot Plate has three independent heating/stirring positions yet only measures 23-5/8 in. L x 10-5/8 in. W (60 x 27 cm).
Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
Fisherbrand™ Single Position Electromagnetic Stirrer
Slim and compact electromagnetic stirrer designed with modern magnetic coil technology. For better mixing, pulse mode is provided in which a stirring bar can rotate clockwise and counter clockwise for every 30 seconds.
Thermo Scientific™ Magnetic Stirrer RT touch series; 220mm top
Stay in control with our low profile digital display stirrers, ideal for routine stirring applications where visible stir speed is required.
Thermo Scientific™ Cimarec+™ Stirring Hotplates Series
Get precise stirring control, exceptional safety and temperature performance for your routine protocols.