missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKB1(phospho S907) & NFKB1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marque: Abnova DP0136.1Set
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the NFKB1 protein, and the other against the specific S907 phosphorylated site of NFKB1 for use in in situ Proximity Ligation Assay.Specifications
NFKB1 | |
Unconjugated | |
DKFZp686C01211/EBP-1/KBF1/MGC54151/NF-kappa-B/NFKB-p105/NFKB-p50/p105/p50 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Phospho-NFKB1 S907 rabbit polyclonal antibody (20 ul) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. NFKB1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 | |
NFKB1 | |
RUO | |
4790 | |
Human | |
Ab Pair |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NFKB1(phospho S907) & NFKB1 Protein Phosphorylation Antibody Pair, Abnova™ > 1 Set; Unconjugated
Spot an opportunity for improvement?Share a Content Correction