Learn More
Klebsiella Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA173177
Description
The PA1-73177 antibody reacts with Klebsiella species. PA1-73177 has been successfully used in Immunofluorescence applications. The PA1-73177 antibody was raised against Klebsiella pneumoniae. Short term (6months) storage is liquid at 4oC. For longer term storage, aliquot and freeze.
Klebsiella are non-motile, rod-shaped, aerobic bacteria that possess a prominent polysaccharide capsule. Some of these bacteria produce an extracellular toxic complex that has been shown to be "e;lethal for and produce extensive lung pathology in mice"e;. It is composed of 63% capsular polysaccharide, 30% lipopolysaccharide, and 7% protein; when introduced to experimental animals in sublethal doses, the animals built up immunization due to antibody production. Klebsiella pneumoniae is one of the most common Gram-negative bacteria seen by physicians worldwide. Pneumonias that are caused by Klebsiella pneumoniae are difficult to control; mortality rates have even been reported as up to 50% after antibiotic treatment. Nitrogen metabolism in Klebsiella aerogenes has also been studied throroughly.
Spécification
Klebsiella | |
Polyclonal | |
Biotin | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Klebsiella pneumoniae | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Certificats
Un numéro de lot est requis pour afficher les résultats sur les certificats. Pour retrouver le numéro de lot sur des commandes passées, utiliser le menu suivi de commande.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.