missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA173141
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The PA1-73141 antibody reacts with Legionella pneumophila. PA1-73141 has been successfully used in ELISA applications. The PA1-73141 antibody was raised against whole cell preparation of Legionella pneumophila; ATCC #33152.
Legionella is found in aquatic environments where its growth is supported by amoebae. In human infections, Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.
Spécification
Legionella | |
Polyclonal | |
HRP | |
L. pneumophila; Legionellosis | |
Whole cell preparation of Legionella pneumophila; ATCC #33152. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1-2 mg/mL | |
PBS with 10mg/mL BSA and 0.002% thimerosal, pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Legionella Polyclonal Antibody, HRP, Invitrogen™ > 1 mL; HRP
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu