Learn More
Annexin VI Mouse, Unlabeled, Clone: 73, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 610301
Description
Annexins are a widely expressed family of Ca2+ and phospholipid-binding proteins. At least ten have been identified in mammalian tissues. They have also been identified in Drosophila, Hydra, and Dictyostelium. Annexin VI is a 70kDa member of the annexin family. While most of the other annexin family members have a four amino acid sequence repeat, Annexin VI displays an eight residue repeat. It is a widely expressed protein, found in lymphocytes, neurons, and many other cell types. Work on annexins has addressed the possible role of Annexin VI in various stages of the endocytic pathway. Some studies suggest that Annexin VI is required for coated-vesicle budding. However, other research directly contradicts this data. Thus, the role of Annexin VI is still under investigation.
Immunofluorescence, Immunoprecipitation, Western Blotting

Spécification
Annexin VI | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Human, Rat | |
IgG1 |
Western Blot | |
73 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.