Learn More
Annexin VI Mouse, Unlabeled, Clone: 73, BD
Mouse Monoclonal Antibody
256.00€ - 504.00€
Spécification
Antigène | Annexin VI |
---|---|
Clone | 73 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15845389
|
BD Biosciences
610300 |
50 μg |
256.00€
1 kit |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
15855389
|
BD Biosciences
610301 |
150 μg | N/A | ||||||
Description
Annexins are a widely expressed family of Ca2+ and phospholipid-binding proteins. At least ten have been identified in mammalian tissues. They have also been identified in Drosophila, Hydra, and Dictyostelium. Annexin VI is a 70kDa member of the annexin family. While most of the other annexin family members have a four amino acid sequence repeat, Annexin VI displays an eight residue repeat. It is a widely expressed protein, found in lymphocytes, neurons, and many other cell types. Work on annexins has addressed the possible role of Annexin VI in various stages of the endocytic pathway. Some studies suggest that Annexin VI is required for coated-vesicle budding. However, other research directly contradicts this data. Thus, the role of Annexin VI is still under investigation.
Immunofluorescence, Immunoprecipitation, Western Blotting
Spécification
Annexin VI | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
73 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.