Learn More
RAB3GAP1 Mouse anti-Human, Mouse, Rat, Clone: 4A3A1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66688-1-IG-150UL
Description
RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.Spécification
RAB3GAP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P69735, Q15042, Q80UJ7 | |
Rab3gap1 | |
RAB3GAP1 Fusion Protein Ag17710 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
4A3A1 | |
Unconjugated | |
RAB3GAP1 | |
DKFZp434A012, KIAA0066, P130, Rab3 GAP, Rab3 GAP p130, RAB3GAP, RAB3GAP1, RAB3GAP130, WARBM1 | |
Mouse | |
Protein G | |
RUO | |
226407, 22930, 304759 | |
-20°C | |
Liquid |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.