missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific™ Product Code.: 15498547

(R)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific™

Product Code. 15498547
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15498547

Brand: Thermo Scientific Alfa Aesar H52055.03

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218608-83-4
Molecular Formula C16H23NO4
Molecular Weight (g/mol) 293.363
MDL Number MFCD01076262
InChI Key MYWZFJXOLAXENE-CYBMUJFWSA-N
Synonym r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, r-3-boc-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, boc-d-?-nva 5-phenyl-oh, n-boc-3-r-amino-5-phenylpentanoic acid, n-tert-butoxycarbonyl-3 r-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonylamino-5-phenyl-pentanoic acid, r-3-tert-butoxycarbonyl amino-5-phenylpentanoicacid, r-3-boc-amino-5-phenylpentanoic acid hplc, 3r-3-2-methylpropan-2-yl oxycarbonylamino-5-phenylpentanoic acidShow MoreShow Less
PubChem CID 7021558
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-phenylpentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)CC(=O)O

Specifications

Quantity 1 g
Formula Weight 293.36
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-5-phenylpentanoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK18W035 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.