missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase I Monoclonal Antibody (10H5), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA123189
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: MCF7.
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
Spécification
DNA Ligase I | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative | |
P18858, P37913 | |
LIG1 | |
Full-length recombinant human DNA Ligase I protein | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
10H5 | |
Unconjugated | |
LIG1 | |
AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1 | |
Mouse | |
Protein G | |
RUO | |
16881, 3978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Suggestions de produits
Customers who viewed this item also viewed.
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
DNA Ligase I Monoclonal Antibody (10H5), Invitrogen™ > 100 μL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu