missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Monoclonal Antibody (3B5), Alexa Fluor™ 700, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MHCD0829
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Alexa Fluor® 700 dye conjugate provides an excellent spectral match to the common long-wavelength excitation sources, with a high extinction coefficient. This antibody recognizes the alpha chain alone as well as the alpha/beta heterodimer.
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry.
Spécification
CD8 | |
Monoclonal | |
Alexa Fluor 700 | |
CD8B | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); Lyt3; Lyt-3; MAL; membrane glycoprotein; membrane protein; OKT8 T-cell antigen; OX-8 membrane antigen; p32; P37; RHACD8-4; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-cell surface glycoprotein Lyt-3; T-cell surface molecule; T-lymphocyte differentiation antigen T8/Leu-2; type I transmembrane glycoprotein | |
Mouse | |
purified | |
ASR | |
925, 926 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
3B5 | |
PBS with BSA and 0.1% sodium azide | |
P01732, P10966 | |
Cd8a, CD8B | |
Human CD8 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
CD8 Monoclonal Antibody (3B5), Alexa Fluor™ 700, Invitrogen™ > 500 μL; Alexa Fluor 700
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu