missing translation for 'onlineSavingsMsg'
Learn More
3-Hydroxy-2-methylpyridine, 99%, Thermo Scientific Chemicals Product Code.: 11421477

3-Hydroxy-2-methylpyridine, 99%, Thermo Scientific Chemicals

Product Code. 11421477
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11421477

Brand: Thermo Scientific Alfa Aesar B24944.14

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-25-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00082538
InChI Key AQSRRZGQRFFFGS-UHFFFAOYSA-N
Synonym 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picolineShow MoreShow Less
PubChem CID 70719
IUPAC Name 2-methylpyridin-3-ol
SMILES CC1=C(C=CC=N1)O

Specifications

Melting Point 166°C to 169°C
Odor Odorless
Quantity 25 g
Beilstein 107937
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-2-methylpyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10144999 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB24944.03,B24944.14,B24944.06
Lot Number10242777 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog NumberB24944.03,B24944.14,B24944.06
Lot Number10242456 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberB24944.03,B24944.14,B24944.06
Lot NumberI11T046 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24944.03,B24944.14,B24944.06
Lot Number10159320 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24944.03,B24944.14,B24944.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Hydroxy-2-methylpyridine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.