Learn More
TRK fused gene Recombinant Rabbit Monoclonal Antibody (SD082-0), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen MA532375
334.68 EUR valable jusqu'au 2025-03-21
Utilisez le code promo "24111" pour bénéficier de cette offre.
Alert:
Pour que la promotion s’applique, le client doit acheter trois fois le même produit au prix catalogue sur la même commande pour avoir une remise de 33.33%. Il n’y a pas de limite d’achat pour un multiple de 3 produits. PCODE: 24111
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.
Spécification
TRK fused gene | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4 | |
Q92734 | |
TFG | |
Synthetic peptide within Human TRK aa 351-400 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot | |
SD082-0 | |
Unconjugated | |
TFG | |
AI173908, HMSNP, protein TFG, SPG57, TF6, TFG, Trk-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene | |
Rabbit | |
Protein A | |
RUO | |
10342, 21787, 360709 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.