missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3C2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22028-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to the phosphoinositide 3-kinase (PI3K) family. PI3-kinases play roles in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. This protein contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases. C2 domains act as calcium-dependent phospholipid binding motifs that mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The PI3-kinase activity of this protein is not sensitive to omolar levels of the inhibitor wortmanin. This protein was shown to be able to be activated by insulin and may be involved in integrin-dependent signaling.Spécification
PIK3C2A | |
Polyclonal | |
Unconjugated | |
PIK3C2A | |
CPK, PI 3 Kinase Class 2A, PI 3-kinase C2alpha, PI3 K C2(ALPHA), PI3 K C2A, PI3K C2 alpha, PIK3C2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18704, 5286 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00443 | |
Pik3c2a | |
PIK3C2A Fusion Protein Ag16894 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-5 of 12
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
PIK3C2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu