missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Endosialin/CD248 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB7535
368.80 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Mouse samples. It is validated for Immunocytochemistry.
Spécification
Endosialin/CD248/TEM1 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q91V98 | |
CD248 | |
Mouse myeloma cell line NS0-derived recombinant mouse Endosialin/CD248 Val13-Arg693 (predicted) Accession # Q91V98 | |
100 μg | |
Primary | |
Detects mouse Endosialin/CD248 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human Endosialin/CD248 is observed. | |
Mouse | |
Purified |
Immunocytochemistry | |
458606 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1, TEM1MGC119478, Tumor endothelial marker 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57124 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-5 of 12
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Mouse Endosialin/CD248 Antibody, R&D Systems™ > 100ug; Unlabeled
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu