Learn More
O&M Halyard Quick Check Sterilization Wrap (H100)
Using Halyard Quick Check wraps is a fast, easy way to ensure instrument sterility is uncompromised.
Marque:  O&M 34192
Code produit. 30274746
  • 238.12€ / 480 pièces
Expédition estimée: 09-05-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
  • Inspect trays easier and faster than ever
  • Superior microbial barrier protection with patented Powerguard technology
  • Cleared for all of the same sterilization systems as Halyard Kimguard One-step Wrap
  • Made from recyclable No. 5 polypropylene fabric
  • Made in the USA
Spécification
Spécification
Sterilization Wrap
Meets NFPA Class 1
Light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs (0 to 3 lb.)
Non stérile
50 x 50 cm (20 x 20 in.)
50 cm
50 cm
Autoclavable
Bleu
SMS Fabric (H100)
Oui
20 in
20 in
Nom du produit
O&M Halyard Quick Check Sterilization Wrap (H100) > H100; 50 x 50 cm (20 x 20 in.)