Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit
Column-free immunomagnetic positive selection of human CD19+ cells for HLA analysis
Marque:  STEMCELL Technologies 17874
Encompass
Code produit. 17137781
  • 1387.50€ / 1 kit
Découvrez plus d'offres spéciales
Description
Description
  • The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells
  • It is also expressed on follicular dendritic cells
  • This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations
Spécification
Spécification
B Cells
60 mL Buffy Coat or Whole Blood
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit