Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Marque: VEE GEE 80906E
Description
- FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial
Spécification
Safety Blue | |
immersion partielle | |
Glass | |
Safety Blue | |
Serialized Glass Thermometer | |
50°C | |
-35°C | |
-35 to +50°C | |
1°C (1.8°F) | |
1.5 cm |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
30.48 cm | |
Immersion/General Purpose (Tapered) | |
5 g | |
122°F | |
-31°F | |
-31 to +122°F | |
±1 subdivision | |
To immersion line |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.