missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-36 beta/IL-1F8 Protein
Recombinant Protein
Marque: Novus Biologicals™ NBP2-34887-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-36 beta/IL-1F8 protein is fully biologically active when compared to standard. The specific activity is determined by its binding ability in a functional ELISA. Immobilized IL-36 beta/IL-1F8 at 1 μg/mL can bind recombinant human IL-1 Rrp2 Fc Chimera with a range of 0.15-5 μg/mL.
Spécification
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. | |
17.7 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
PAGE, Bioactivity | |
27177 | |
100 μg | |
Less than 1 EU/ug of IL-36 beta/IL-1F8 as determined by LAL method. | |
IL36B | |
Biologically Active Proteins | |
IL-36 beta/IL-1F8 |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Novus Biologicals™ Recombinant Human IL-36 beta/IL-1F8 Protein > 100 μg
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu