missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Potassium iodide is an ionic compound that has been used as a medication (particularly in response to exposure to radioactive iodine). It is a mild reducing agent that is easily oxidized. It is used in iodine isolation, in redox titrations and as a source of iodide in organic synthesis. It may also act as a nucleophilic catalyst for the alkylation of alkyl chlorides, bromides or mesylates.
Specifications
Specifications
Quantity | 500 mL |
Formula Weight | 166.01g/mol |
Concentration or Composition (by Analyte or Components) | 10% |
Density | 1.07g/mL |
Safety and Handling

Product Identifier
- Potassium iodide 10% solution
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : KI/H2O
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Iodide, 10% Solution, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction