Learn More
NTRK2 Mouse anti-Human, Clone: OTI2E1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies CF500386

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TrkB is a receptor tyrosine kinase of the Trk family. Receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3, -4 and -5 but not nerve growth factor (NGF). TrkB is involved in the development and maintece of the nervous system. Moreover, Trk B is a tyrosine kinase gene highly related to Trk A. Trk B expression is confined to tissues within the central and peripheral nervous systems. The brain-derived neurotrophic factor (BDNF) and NT-3, but not NGF, can induce rapid phosphorylation on tyrosine of Trk B gp145, one of the receptors encoded by Trk B, although BDNF elicits a response at least two orders of magnitude greater than NT-3. TrkB is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signaling through TrkB leads to cell differentiation. Mutations in the TrkB gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for the TrkB gene.Spécification
NTRK2 | |
Monoclonal | |
Unconjugated | |
NTRK2 | |
EIEE58, GP145-TrkB, OBHD, TRKB, trk-B | |
Mouse | |
Affinity Chromatography | |
RUO | |
4915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16620 | |
NTRK2 | |
Full-length protein expressed in 293T cell transfected with human NTRK2 expression vector | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.