missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™ Product Code.: 15428555

Ethyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15428555
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15428555

Brand: Thermo Scientific Alfa Aesar H33631.06

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35978-33-7
Molecular Formula C13H12FNO2S
Molecular Weight (g/mol) 265.302
MDL Number MFCD00239291
InChI Key CJMZLCRLBNZJQR-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-fluorophenyl thiophene-3-carboxylate, 2-amino-4-4-fluoro-phenyl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-4-fluorophenyl-3-thiophenecarboxylate, ethyl2-amino-4-4-fluorophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-fluophenyl thiophene-3-carboxylate, maybridge1_000485, 3-thiophenecarboxylicacid, 2-amino-4-4-fluorophenyl-, ethyl ester
PubChem CID 613091
IUPAC Name ethyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)F)N

Specifications

Melting Point 87°C to 94°C
Quantity 5 g
Formula Weight 265.3
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-amino-4-(4-fluorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.