Learn More
Novus Biologicals™ CD63 Recombinant Protein Antigen
Recombinant Protein
Marque: Novus Biologicals™ NBP3-21363PEP
Description
Spécification
Antibody Competition | |
967 | |
100 μL | |
CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-21363. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL | |
>80% by SDS-PAGE and Coomassie blue staining |
PBS and 1 M Urea, pH 7.4. | |
MolecularWeight-theroretical: 20 kDa | |
Store at -20°C. Avoid freeze/thaw cycles. | |
CD63 | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
CD63 |
Suggestions de produits
Customers who viewed this item also viewed.