Learn More
CD144 Rat anti-Mouse, PE, Clone: 11D4.1, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 562243
Description
The 11D4.1 antibody monoclonal antibody specifically binds to mouse CD144, also known as VE-cadherin. CD144 is a type I transmembrane protein and is a member of the cadherin superfamily. VE-cadherin is an endothelial cell-specific, homophilic adhesion molecule. It is concentrated at interendothelial cells contacts and is thought to be involved in the maintenance of cell layer integrity. In vitro and in vivo studies indicate that the 11D.4 mAb interferes with VE-cadherin-mediated intercellular adhesion.

Spécification
CD144 | |
Monoclonal | |
0.2mg/mL | |
Cdh5; Cadherin-5; CADH5; VE-cadherin; Vascular endothelial cadherin; 7B4 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
11D4.1 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse VE-Cadherin-Ig Fusion | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.