missing translation for 'onlineSavingsMsg'
Learn More
3-Cyclopropylpropargyl diethyl acetal, 97%, Thermo Scientific™ Product Code.: 10617094

3-Cyclopropylpropargyl diethyl acetal, 97%, Thermo Scientific™

Product Code. 10617094
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10617094

Brand: Thermo Scientific Acros 391650050

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 146195-69-9
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD12912455
InChI Key WOSJTIAXBXVXLF-UHFFFAOYSA-N
Synonym 3-cyclopropylpropiolaldehyde diethyl acetal, 3,3-diethoxyprop-1-yn-1-yl cyclopropane, 3-cyclopropylpropargyl diethyl acetal, 3-cyclopropylpropargyl aldehyde diethyl acetal
PubChem CID 42580279
IUPAC Name 3,3-diethoxyprop-1-ynylcyclopropane
SMILES CCOC(C#CC1CC1)OCC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9700g/mL
Boiling Point 236°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5CCCH(OCH2CH3)2
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.97
Formula Weight 168.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Cyclopropylpropargyl diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyclopropylpropargyl diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Cyclopropylpropargyl diethyl acetal, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.