missing translation for 'onlineSavingsMsg'
Learn More
2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™ Product Code.: 10605165

2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10605165
10 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10605165

Brand: Thermo Scientific Maybridge CC34701EA

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21278-86-4
Molecular Formula C9H5N2O2S
Molecular Weight (g/mol) 205.21
MDL Number MFCD00171745
InChI Key COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonym 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
IUPAC Name 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid
SMILES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Quantity 10 g
Formula Weight 206.22
Percent Purity 95%
Chemical Name or Material 2-(4-pyridyl)-1, 3-thiazole-4-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.