Inorganic Gadolinium Compounds
Nitrate de gadolinium(III) hexahydraté, 99,9 %, (base métallique à l’état de traces), Thermo Scientific Chemicals
CAS: 19598-90-4 Formule moléculaire: GdN3O9·6H2O Poids moléculaire (g/mol): 451.4 Numéro MDL: MFCD00149728 Clé InChI: XWFVFZQEDMDSET-UHFFFAOYSA-N Synonyme: gadolinium iii nitrate hexahydrate trace metals basis, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium nitrate, hexahydrate,, gadolinium 3+ trinitrate hexahydrate, gd.3no3.6h2o, gadoliniumnitratehexahydrate, nitric acid, gadolinium 3+ salt, hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, gadolinium nitrate hexahydrate, gadolinium iii nitrate hexahydrate CID PubChem: 209258 Nom IUPAC: gadolinium(3+);trinitrate;hexahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]
Chlorure de gadolinium(III) hexahydraté, Reacton™, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 13450-84-5 Formule moléculaire: Cl3GdH12O6 Poids moléculaire (g/mol): 371.69 Numéro MDL: MFCD00149732 Clé InChI: PNYPSKHTTCTAMD-UHFFFAOYSA-K Synonyme: gadolinium iii chloride hexahydrate, reacton reo, gadolinium chloride, hexahydrate,, gadolinium iii chloridehexahydrate, gadolinium 3+ trichloride hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride, hydrate 1:6, unii-261ex557hi, gadolinium trichloride hexahydrate, gadolinium chloride hexahydrate CID PubChem: 197915 SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
Oxyde de gadolinium(III), 99,9 % (REO), Thermo Scientific Chemicals, REacton™
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Hydrate de bromure de gadolinium(III), Reacton,™,99,99 % (REO), Thermo Scientific Chemicals
CAS: 30010-20-9 Formule moléculaire: Br3Gd Poids moléculaire (g/mol): 396.96 Numéro MDL: MFCD00049741 Clé InChI: KGOKDPWKDBWITQ-UHFFFAOYSA-K Nom IUPAC: gadolinium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Gd+3]
Le pentane2,4dione de gadolinium(III) hydraté, REacton™, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 64438-54-6 Formule moléculaire: C15H21GdO6 Poids moléculaire (g/mol): 454.58 Numéro MDL: MFCD00013493 Clé InChI: PJCXQIZIMGZZIT-UHFFFAOYSA-N Synonyme: r reo, gadolinium iii 2,4-pentanedionate hydrate, reacton CID PubChem: 131675701 Nom IUPAC: gadolinium(3+) ; (Z)-4-oxopent-2-en-2-olate ; hydrate SMILES: [Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Chlorure de gadolinium(III), ultra sec, 99,99 %, Thermo Scientific Chemicals
CAS: 10138-52-0 Formule moléculaire: Cl3Gd Poids moléculaire (g/mol): 263.6 Numéro MDL: MFCD00011024 Clé InChI: MEANOSLIBWSCIT-UHFFFAOYSA-K Synonyme: gadolinium iii chloride, anhydrous, ?gdcl3, gadolinium iii chloride, ultra dry, wln: gd g3, gadolinium 3+ chloride, gadolinium iii chloride, gdcl3, gadolinium chloride gdcl3, gadolinium trichloride, gadolinium chloride CID PubChem: 61486 ChEBI: CHEBI:37288 Nom IUPAC: trichlorogadolinium SMILES: Cl[Gd](Cl)Cl
Oxyde de gadolinium(III), 99,9 % (REO), Thermo Scientific Chemicals, REacton™
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Chlorure de gadolinium(III) hexahydraté, agrégats cristallins blancs, REacton™, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 13450-84-5 Formule moléculaire: Cl3GdH12O6 Poids moléculaire (g/mol): 371.69 Numéro MDL: MFCD00149732 Clé InChI: PNYPSKHTTCTAMD-UHFFFAOYSA-K Synonyme: gadolinium iii chloride hexahydrate, reacton reo, gadolinium chloride, hexahydrate,, gadolinium iii chloridehexahydrate, gadolinium 3+ trichloride hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride, hydrate 1:6, unii-261ex557hi, gadolinium trichloride hexahydrate, gadolinium chloride hexahydrate CID PubChem: 197915 SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
Nitrate de gadolinium(III) hydraté, REacton™, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 94219-55-3 Formule moléculaire: GdN3O9 Poids moléculaire (g/mol): 343.26 Numéro MDL: MFCD00011023 Clé InChI: MWFSXYMZCVAQCC-UHFFFAOYSA-N Nom IUPAC: gadolinium(3+) trinitrate SMILES: [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Fluorure de gadolinium(III), anhydre, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 13765-26-9 Formule moléculaire: F3Gd Poids moléculaire (g/mol): 214.25 Numéro MDL: MFCD00016113 Clé InChI: TYIZUJNEZNBXRS-UHFFFAOYSA-K Nom IUPAC: gadolinium(3+) trifluoride SMILES: [F-].[F-].[F-].[Gd+3]
Oxyde de gadolinium(III), poudre ou granulés blancs, REacton™, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Oxyde de gadolinium(III), 99,9 % (REO), Thermo Scientific Chemicals
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 Nom IUPAC: gadolinium(+)3+) ; oxygène(2-) SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Nitrate de gadolinium(III) pentahydraté, 99,9 %, Thermo Scientific Chemicals
CAS: 52788-53-1 Formule moléculaire: GdN3O9·5H2O Poids moléculaire (g/mol): 433.33 Numéro MDL: MFCD00149729
Carbonate de gadolinium(III) hydraté, 99 % (REO), Thermo Scientific Chemicals
CAS: 38245-36-2 Formule moléculaire: C3Gd2O9 Poids moléculaire (g/mol): 494.52 Numéro MDL: MFCD00150240 Clé InChI: RQXZRSYWGRRGCD-UHFFFAOYSA-H Synonyme: gadolinium iii carbonate hydrate-gd reo puratrem, gadolinium iii carbonate hydrate, reacton?, digadolinium 3+ hydrate tricarbonate, gadolinium carbonate-water 2/3/1, 3co3.2gd.h2o, acmc-1aeff, gadolinium iii carbonate hydrate, gadolinium carbonate hydrate CID PubChem: 16212943 SMILES: [Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Oxyde de gadolinium(III), 99,9 % (base de métaux à l’état de traces), Thermo Scientific Chemicals
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Hydrate d’hydroxyde de gadolinium(III), 99,9 % (REO), Thermo Scientific Chemicals
CAS: 100634-91-1 Formule moléculaire: GdH3O3 Poids moléculaire (g/mol): 208.27 Numéro MDL: MFCD00049930 Clé InChI: ILCLBMDYDXDUJO-UHFFFAOYSA-K Nom IUPAC: gadolinium(3+) trihydroxide SMILES: [OH-].[OH-].[OH-].[Gd+3]
Bromure de gadolinium(III), ultra sec, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 13818-75-2 Numéro MDL: MFCD00049741
Perchlorate de gadolinium(III) hexahydraté, 50 % w/w soln. aq., qualité réactif, Thermo Scientific Chemicals
CAS: 14017-52-8 Numéro MDL: MFCD00150239 Synonyme: gadolinium iii perchlorate solution, 40 wt. % in h2o, gadolinium perchlorate, crystalline hydrate,, gadolinium 3+ triperchlorate, gadolinium iii perchlorate solution, acmc-20aku7, gadolinium iii perchlorate, gadolinium perchlorate
Hydrate d’acétate de gadolinium(III), cristaux blancs, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 100587-93-7 Formule moléculaire: C6H9GdO6 Poids moléculaire (g/mol): 334.38 Numéro MDL: MFCD00150116 Clé InChI: LYQGMALGKYWNIU-UHFFFAOYSA-K Nom IUPAC: gadolinium(3+) triacetate SMILES: [Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Borure de gadolinium, 99,5 % (base métallique), Thermo Scientific Chemicals
CAS: 12008-06-9 Formule moléculaire: B6Gd Poids moléculaire (g/mol): 222.11 Numéro MDL: MFCD00049942 Clé InChI: FYKPSYVUFHLGOY-UHFFFAOYSA-N Synonyme: gadolinium 2+ ion pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, gadolinium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide, gadolinium boride, gdb6, gadolinium boride CID PubChem: 25021694 Nom IUPAC: Gadolinium(2+) pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide SMILES: [Gd++].B12B3[B-]11B4B1[B-]234
Iodure de gadolinium(III), ultra-sec, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 13572-98-0 Formule moléculaire: GdI3 Poids moléculaire (g/mol): 537.96 Numéro MDL: MFCD00016114 Clé InChI: IZZTUGMCLUGNPM-UHFFFAOYSA-K Nom IUPAC: Triiodure de gadolinium(3+) SMILES: [I-].[I-].[I-].[Gd+3]
Sulfate de gadolinium(III) octahydraté, REacton™, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 13450-87-8 Formule moléculaire: Gd2H16O20S3 Poids moléculaire (g/mol): 746.788 Numéro MDL: MFCD00149734 Clé InChI: JBFCVGZGWFIPNK-UHFFFAOYSA-H Synonyme: gadolinium iii sulfate octahydrate trace metals basis, gadolinium iii sulfate octahydrate, reacton, gadolinium sulfate-water 2/3/8, gadolinium sulfate, gadolinium sulfuric acid octahydrate, gadolinium sulfate octahydrate, gadolinium iii sulfate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate octahydrate CID PubChem: 16211473 Nom IUPAC: gadolinium(3+);trisulfate;octahydrate SMILES: O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]
Hydrate de carbonate de gadolinium(III), REacton™, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 38245-36-2 Formule moléculaire: C3Gd2O9 Poids moléculaire (g/mol): 494.52 Numéro MDL: MFCD00150240 Clé InChI: RQXZRSYWGRRGCD-UHFFFAOYSA-H Synonyme: gadolinium iii carbonate hydrate-gd reo puratrem, gadolinium iii carbonate hydrate, reacton?, digadolinium 3+ hydrate tricarbonate, gadolinium carbonate-water 2/3/1, 3co3.2gd.h2o, acmc-1aeff, gadolinium iii carbonate hydrate, gadolinium carbonate hydrate CID PubChem: 16212943 Nom IUPAC: gadolinium(3+) ; tricarbonate ; hydrate SMILES: [Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Oxyde de gadolinium(III), nanopoudre, + de 99,99 % (REO), Thermo Scientific Chemicals
CAS: 12064-62-9 Formule moléculaire: Gd2O3 Poids moléculaire (g/mol): 362.50 Numéro MDL: MFCD00011026 Clé InChI: CMIHHWBVHJVIGI-UHFFFAOYSA-N Synonyme: digadolinium 3+ ion trioxidandiide, gadolinium 3+ ; oxygen 2-, unii-5480d0nhlj, gadolinium 3+ oxide, gadolinium sesquioxide, digadolinium trioxide, gadolinium trioxide, gadolinium iii oxide, gadolinium oxide, gadolinia CID PubChem: 159427 Nom IUPAC: gadolinium(+)3+) ; oxygène(2-) SMILES: [O--].[O--].[O--].[Gd+3].[Gd+3]
Fluorure de gadolinium(III) anhydre, Reacton,™, 99,99 % (REO), Thermo Scientific Chemicals
CAS: 13765-26-9 Formule moléculaire: F3Gd Poids moléculaire (g/mol): 214.25 Numéro MDL: MFCD00016113 Clé InChI: TYIZUJNEZNBXRS-UHFFFAOYSA-K Nom IUPAC: gadolinium(3+) trifluoride SMILES: [F-].[F-].[F-].[Gd+3]
Bromure de gadolinium(III), ultra sec, 99,99 % (base métallique), Thermo Scientific Chemicals
CAS: 13818-75-2 Formule moléculaire: GdBr3 Numéro MDL: MFCD00049741
Chlorure de gadolinium(III), 99,9 %, (métaux à l’état de traces), anhydre, Thermo Scientific Chemicals
CAS: 10138-52-0 Formule moléculaire: Cl3Gd Poids moléculaire (g/mol): 263.61 Numéro MDL: MFCD00011024 Clé InChI: MEANOSLIBWSCIT-UHFFFAOYSA-K Synonyme: gadolinium iii chloride, anhydrous, ?gdcl3, gadolinium iii chloride, ultra dry, wln: gd g3, gadolinium 3+ chloride, gadolinium iii chloride, gdcl3, gadolinium chloride gdcl3, gadolinium trichloride, gadolinium chloride CID PubChem: 61486 ChEBI: CHEBI:37288 Nom IUPAC: trichlorogadolinium SMILES: Cl[Gd](Cl)Cl
Nitrure de gadolinium(III), 99,5 % (REO), Thermo Scientific Chemicals
CAS: 25764-15-2 Formule moléculaire: GdN Poids moléculaire (g/mol): 171.26 Numéro MDL: MFCD00049943 Clé InChI: FLATXDRVRRDFBZ-UHFFFAOYSA-N Synonyme: gadoliniumylidyneamine, gadolinium iii nitride, gadolinium nitride CID PubChem: 117632 Nom IUPAC: gadolinioamine SMILES: N#[Gd]
Nitrate de gadolinium(III) hydraté, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 94219-55-3 Formule moléculaire: GdN3O9 Poids moléculaire (g/mol): 343.26 Numéro MDL: MFCD00011023 Clé InChI: MWFSXYMZCVAQCC-UHFFFAOYSA-N Nom IUPAC: gadolinium(3+) trinitrate SMILES: [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Hydrate d’oxalate de gadolinium(III), 99,9 % (REO), Thermo Scientific™
CAS: 100655-00-3 Formule moléculaire: C6H8Gd2O13 Poids moléculaire (g/mol): 602.62 Numéro MDL: MFCD00150145 Clé InChI: FEIDMMDEDUAFJQ-UHFFFAOYSA-N Synonyme: oxalic acid-gadolinium-water 3/2/1 CID PubChem: 92020445 Nom IUPAC: gadolinium;acide oxalique;hydrate SMILES: O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O