Learn More
VEE GEE Thermometer, Encapsulated in FEP
The encapsulated glass thermometer is designed to safeguard against broken glass and liquid. It contains a biodegradable Safety Blue fill, offering mercury-like properties minus the associated risks of contamination and hazards.
Marque: VEE GEE 80904E
Description
- FEP encapsulation- in the event of breakage the FEP coating contains the broken glass and liquid, preventing injury or contamination
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial
Spécification
Safety Blue | |
immersion partielle | |
Glass | |
Safety Blue | |
Serialized Glass Thermometer | |
104.4°C | |
-1.1°C | |
-1.1 to 104.4°C | |
1.1°C (2°F) | |
1.65 cm |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
39 cm | |
Immersion/General Purpose (Tapered) | |
14 g | |
220°F | |
30°F | |
30 to 220°F | |
±1 subdivision | |
To immersion line |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.