Learn More
FGF-23 Antibody (FGF23/132), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-20779AF594
Description
FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
FGF-23 | |
Monoclonal | |
Alexa Fluor 594 | |
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
Recombinant full-length human FGF-23 protein | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
8074 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
FGF23/132 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.