Learn More
VEE GEE Dial Thermometer
Dial thermometers of varying sizes based on personal preferences.
Marque: VEE GEE 82160-6
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- The dial thermometer shows the temperature using a metal stem and a circular scale.
- It doesn't need batteries, it's tough, and you can change the readings by using the calibration screw on the back.
Spécification
7.6 cm Diameter | |
1/2 in. NPT male thread on the back for threading into a pipeline and an adjustment screw for field calibration. | |
15.2 cm | |
Serialized Glass Thermometer | |
160°F | |
-40°F | |
-40 to +160°F | |
±1 subdivision | |
15.2 cm |
Food & beverage quality control, general lab use, and asphalt/concrete testing | |
12 in. | |
Metal | |
Case is hermetically sealed to prevent lens fogging | |
71.1°C | |
-40°C | |
-40 to +71.1°C | |
2°F |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Nom du produit
VEE GEE Dial Thermometer > Temperature Range: -40 to +160°F, Display Dimensions: 7.6 cm Diameter, Immersion Length: 15.2 cm, Probe Length: 12 in.
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu