Learn More
Clifton™ Advanced Digital Water Bath Europe
Advanced digital water bath with PID temperature control. Controls feature auto alarm setting, illuminated on/off switch and over temperature indicators. Supplied with a gated drain tap.
Marque: Clifton™ NE2-28DEU
Description
- Advanced digital water bath
- 28 L with PID temperature control
- Controls feature auto alarm setting, illuminated on/off switch and over temperature indicators
- Supplied with a gated drain tap
Spécification
Non recommandé | |
Digital Controller | |
Ambient +5°C to 100°C | |
240V |
28 L | |
LD-28, GL1-22AF, SL1-22H, SL1-22, GL1-22, BSK-22, BA-12, | |
Water Bath | |
230 V |