Learn More
Dupont™ Tyvek™ IsoClean™ Lab Coats Code produit.: 15874158
Dupont™ Tyvek™ IsoClean™ Lab Coats
Code produit. 15874158 Tous les produits DuPont Produits
Small
Click to view available options
Dimensions:
Small
X-Large
2X-Large
3X-Large
Code produit. 15874158
Marque: Dupont™ D13472742
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
DuPont™ Tyvek™ IsoClean™ labcoats deliver an ideal balance of protection, durability and comfort. These coats are manufactured from a high density polyethylene using a patented flash spinning process.
  • With bound neck and internal seams
  • Available in sizes from Small to 3X
  • Covered elastication at wrists
  • Front snap closure for easy donning and doffing
  • Not clean-processed and not sterilized
  • Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splashes
  • Breathable and exceptionally low linting
  • Manufactured in a controlled environment
  • Typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry and electronics industries as well as in other critical or controlled environments
  • 30 per box, bulk packed
TRUSTED_SUSTAINABILITY
Spécification
Type de produit Controlled Environments Lab Coat
Dimensions Small
Matériau Polyethylene
Couleur Blanc
Jetable Single-use
Stérilité Non stérile
Correction du contenu d'un produit
Nom du produit
Dupont™ Tyvek™ IsoClean™ Lab Coats >