Learn More
Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate, tech. 85%, Thermo Scientific Chemicals
Description
Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate is useful for the preparation of water-soluble organometal catalysts, which finds application in the hydroformylation of propene. It acts as a Lewis base, which is stronger than triphenylphosphine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 g |
Beilstein | 3584807 |
Solubility Information | Soluble in water. |
SMILES | [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC(=CC=C1)S([O-])(=O)=O)C1=CC(=CC=C1)S([O-])(=O)=O |
Formula Weight | 568.41 (Anhydrous) |
Percent Purity | 85% |
Grade | Technical |
Physical Form | Crystalline |
Chemical Name or Material | Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.