Learn More
Siglec-8 Mouse anti-Human, BV750, Clone: 837535, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 747869
530.72 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
Siglec-8 Monoclonal antibody specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry
Spécification
Siglec-8 | |
Monoclonal | |
Brilliant Violet 750 | |
CDw329; SAF-2; SAF2; SIGLEC-8; SIGLEC8 | |
Human Siglec-8 Recombinant Protein | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
837535 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.