missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine, 97+%

Product Code. 15485486
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15485486

Brand: Thermo Scientific Alfa Aesar H60122.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

CAS: 913196-43-7 | C20H20NP | 305.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913196-43-7
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.36
MDL Number MFCD17013987
InChI Key KPFJIPHGQGHIMM-XISACWJONA-N
Synonym s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
PubChem CID 57514668
IUPAC Name (1S)-1-(2-diphenylphosphanylphenyl)ethanamine
SMILES C[C@H](N)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 76°C to 81°C
Quantity 1 g
Sensitivity Air Sensitive
Optical Rotation −55° (c=0.5 in Chloroform)
Formula Weight 305.35
Percent Purity ≥97%
Chemical Name or Material (S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.