missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, 8M aq. soln., RNAse free

Product Code. 15455159
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
This item is not returnable. View return policy

Product Code. 15455159

Brand: Thermo Scientific Alfa Aesar J62856.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium Acetate is a potassium salt employed to replenish electrolytes, for restoration of water-electrolyte balance, as well as a urinary and systemic alkalizer. Potassium Acetate can be administered orally or by intravenous infusion. Formerly, It was employed in diuretics and expectorants. A potassium source employed in DNA and protein purification. Some other applications includes as buffering agent, neutralizer, preservative, color retention agent(protect natural color of animals and plants).

Solubility
Soluble in water (98.2 g/L completely soluble) at 25°C, alcohol, methanol, and ammonia. Insoluble in acetone, and ether.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Linear Formula CH3CO2K
Solubility Information Soluble in water (98.2g/L completely soluble) at 25°C,alcohol,methanol,and ammonia. Insoluble in acetone,and ether.
SMILES CC(=O)[O-].[K+]
Molecular Weight (g/mol) 98.142
Formula Weight 98.15
Concentration or Composition (by Analyte or Components) 8M aq. soln.
Chemical Name or Material Potassium acetate, RNAse free

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.