missing translation for 'onlineSavingsMsg'
Learn More
Magnesium gluconate hydrate, Thermo Scientific Chemicals Product Code.: 16524294

Magnesium gluconate hydrate, Thermo Scientific Chemicals

Product Code. 16524294
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 16524294

Brand: Thermo Scientific Acros 464831000

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336879-53-9
Molecular Formula C12H24MgO15
Molecular Weight (g/mol) 432.61
MDL Number MFCD00150971
InChI Key IAKLPCRFBAZVRW-XRDLMGPZSA-L
IUPAC Name magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate
SMILES O.[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Melting Point 169.7°C to 171.9°C
Quantity 100 g
Solubility Information Solubility aq. soln.: 9.6 g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 414.61
Chemical Name or Material Magnesium gluconate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450029 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number464831000,464830250
Lot NumberA0450031 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog Number464831000,464830250
Lot NumberA0440258 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog Number464831000,464830250
Lot NumberA0410594 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog Number464831000,464830250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium gluconate hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.