Learn More
Technical Service Consultants™ Hygiene Sponge Sampling Kit
Use Technical Service Consultants™ Hygiene Sponge Sampling Kit enables sampling in difficult to reach places, such as crevices and corners. It comes pre-dosed with a neutralizing buffer, swab heads, shaft materials,,tubes for convenient storage and handling and medical-grade plastics for assured quality. .
Marque: Technical Service Consultants™ TS/15-B.
Description
- M40 Compliant
- 2-year shelf life
- Tamper-evident packaging
- Swab heads and shaft materials
- Rackable tubes
- Medical grade plastics
Spécification
Hygiene Sponge Sampling Kits | |
Swab heads and shaft materials, rackable tubes, medical grade plastics | |
4 to 25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container |
Stérile | |
200Pack | |
Viscose |