missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody (rERBB2/9401), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-23594-20ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
ErbB2/Her2 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 μg/mL | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Recombinant fragment (around aa1155-1255) of human ErbB2/Her2 protein (exact sequence is proprietary) | |
20 μg | |
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
2064 | |
Store at 4°C. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
rERBB2/9401 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
ErbB2/Her2 Antibody (rERBB2/9401), Novus Biologicals™ > 20 μg, Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu