Learn More
TFA Dostmann Digital Indoor/Outdoor Thermometer with Alarm
The digital indoor-outdoor thermometer with alarm can monitor the peakvalues over a selected period of time due to its max.-min. memory. Also a temperature alarm can be set. The large measuring range and waterproof sensor allows the use for temperature control of the refrigerator and freezer.
Marque: TFA Dostmann 30.1024
Description
This functional thermometer is ideal for simultaneous measurement of indoor and outdoor temperatures. Controlling the room temperature is an important factor for a comfortable and healthy living environment and it helps save energy. Even one degree less temperature can bring up to 6% reduction in heating costs. Placed at the window, the sensor cable is simply led outside to measure the outside temperature. The max.-min. memory with manual reset allows the observation of the peak values over a self-selected period of time. A temperature alarm can be set to emit an alarm signal during frost or extremely high temperatures. The application possibilities of thermometers with sensor cables are manifold and not only limited to the outdoor temperature. Due to the large measuring range, the waterproof measuring sensor can also be used for temperature control of the refrigerator and freezer or for measuring the water temperature in the aquarium.
- Display of indoor-outdoor temperatures
- Outdoor temperature via cable
- Adjustable alarm value for outdoor temperature
- Also for the monitoring of freezer / aquarium
- Max.-min.-function
Spécification
Plastic | |
Oui | |
LCD |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté