missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 97%, Thermo Scientific™

Product Code. 10449120 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
10g; Glass bottle
This item is not returnable. View return policy

Product Code. 10449120

Brand: Thermo Scientific Acros 170230100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4815-30-9
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H15NO4S
MDL Number MFCD00005450
Quantity 10g
Synonym diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate, diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate, diethyl 5-amino-3-methyl-thiophene-2,4-dicarboxylate, diethyl5-amino-3-methylthiophene-2,4-dicarboxylate, 2,4-thiophenedicarboxylic acid, 5-amino-3-methyl-, diethyl ester, 2-amino-3,5-bis ethoxycarbonyl-4-methylthiophene, 5-amino-3-methyl-thiophene-2,4-dicarboxylic acid diethyl ester, 2-amino-4-methyl-3,5-bis ethoxycarbonyl thiophene, diethyl 5-amino-3-methyl-2,4-thiophene dicarboxylate
InChI Key DGVXLHAJVRRLGV-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(C)C(C(=O)OCC)=C(N)S1
IUPAC Name diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate
Molecular Weight (g/mol) 257.30
PubChem CID 78537
Formula Weight 257.29
Percent Purity 97%
Chemical Name or Material Diethyl 5-amino-3-methyl-2, 4-thiophenedicarboxylate, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.