missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX6 Monoclonal Antibody (8G6), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA549268
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Spécification
DDX6 | |
Monoclonal | |
500 μg/mL | |
PBS with 4MG trehalose and no preservative | |
P26196, P54823 | |
DDX6 | |
E.coli-derived human DDX6 recombinant protein (Position: Q56-P483). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
8G6 | |
Unconjugated | |
DDX6 | |
1110001P04Rik; ATP-dependent RNA helicase p54; DDX6; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); DEAD-box helicase 6; DEAD-box protein 6; E230023J21Rik; HLR2; mRCK/P54; Oncogene RCK; oncogene RCK homolog; p54; probable ATP-dependent RNA helicase DDX6; RCK; RGD1564560 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
13209, 1656, 500988 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2b |
Suggestions de produits
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
DDX6 Monoclonal Antibody (8G6), Invitrogen™ > 100 μg; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu