Learn More
CD27 Mouse anti-Human , PE, Clone: O323, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566945
65.27 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry
Spécification
CD27 | |
Monoclonal | |
PE | |
CD27 antigen; TNFRSF7; S152; T14; Tp55 | |
Not Reported | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
O323 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.