Learn More
ALDH1L1 Antibody (ALDH1L1/7959), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-24222MFV450
564.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Description
ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
ALDH1L1 | |
Monoclonal | |
mFluor Violet 450 SE | |
10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
Recombinant fragment of human ALDH1L1 protein (exact sequence is proprietary) | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction | |
10840 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
ALDH1L1/7959 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Suggestions de produits
Customers who viewed this item also viewed.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.