Learn More
4-méthoxy-N-méthylbenzylamine, 97 %, Thermo Scientific™
51.20€ - 254.00€
Identifiants chimiques
CAS | 702-24-9 |
---|---|
Formule moléculaire | C9H13NO |
Poids moléculaire (g/mol) | 151.209 |
Numéro MDL | MFCD04115407 |
Clé InChI | AIJFPNKGGAPZFJ-UHFFFAOYSA-N |
Synonyme | 4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine |
CID PubChem | 485407 |
Nom IUPAC | 1-(4-méthoxyphényl)-N-méthylméthanamine |
SMILES | CNCC1=CC=C(C=C1)OC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
702-24-9 | |
151.209 | |
AIJFPNKGGAPZFJ-UHFFFAOYSA-N | |
485407 | |
CNCC1=CC=C(C=C1)OC |
C9H13NO | |
MFCD04115407 | |
4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine | |
1-(4-méthoxyphényl)-N-méthylméthanamine |
Spécification
4-Methoxy-N-methylbenzylamine | |
C9H13NO | |
UN2735 | |
AIJFPNKGGAPZFJ-UHFFFAOYSA-N | |
1-(4-méthoxyphényl)-N-méthylméthanamine | |
485407 | |
97% |
702-24-9 | |
MFCD04115407 | |
4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine | |
CNCC1=CC=C(C=C1)OC | |
151.209 | |
151.21 |
Sécurité et traitement
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H361
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon