missing translation for 'onlineSavingsMsg'
Learn More

4-Carbamoyl-3-chlorobenzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15409397
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15409397

Brand: Thermo Scientific Alfa Aesar H52641.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-52-5
Molecular Formula C7H7BClNO3
Molecular Weight (g/mol) 199.40
MDL Number MFCD07363777
InChI Key ZARZNNIZDSRQFA-UHFFFAOYSA-N
Synonym 4-carbamoyl-3-chlorophenyl boronic acid, boronic acid,b-4-aminocarbonyl-3-chlorophenyl, 4-aminocarbonyl-3-chloro benzeneboronic acid, acmc-209q3a, 4-carbamoyl-3-chlorophenyboronic acid, 4-carbamoyl-3-chlorophenyl boronicacid, 4-aminocarbonyl-3-chlorophenyl boronic acid, 4-aminocarbonyl-3-chlorobenzeneboronic acid, 3-chloro-4-carbamoylphenyl boronic acid
PubChem CID 44119639
IUPAC Name (4-carbamoyl-3-chlorophenyl)boronic acid
SMILES NC(=O)C1=C(Cl)C=C(C=C1)B(O)O

Specifications

Melting Point 152°C to 158°C
Quantity 1 g
Formula Weight 199.4
Percent Purity 96%
Chemical Name or Material 4-Carbamoyl-3-chlorobenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.