missing translation for 'onlineSavingsMsg'
Learn More

3-[3-Chloro-5-(trifluoromethoxy)phenyl]propionic acid, 94%, Thermo Scientific™

Product Code. 15477865
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15477865

Brand: Thermo Scientific Alfa Aesar H31679.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-74-1
Molecular Formula C10H8ClF3O3
Molecular Weight (g/mol) 268.616
MDL Number MFCD09025398
InChI Key OOXKNRUBGDQVIF-UHFFFAOYSA-N
Synonym 3-3-chloro-5-trifluoromethoxy phenyl propionic acid, 3-3-chloro-5-trifluoromethoxy phenyl propanoic acid, 3-3-chloro-5-trifluoromethoxy phenyl propanoic a
PubChem CID 46737566
IUPAC Name 3-[3-chloro-5-(trifluoromethoxy)phenyl]propanoic acid
SMILES C1=C(C=C(C=C1OC(F)(F)F)Cl)CCC(=O)O

Specifications

Refractive Index 1.4755
Quantity 1 g
Formula Weight 268.62
Percent Purity 94%
Chemical Name or Material 3-[3-Chloro-5-(trifluoromethoxy)phenyl]propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.