missing translation for 'onlineSavingsMsg'
Learn More
2-Iodobenzotrifluoride, 99%, Thermo Scientific Chemicals Product Code.: 11457657

2-Iodobenzotrifluoride, 99%, Thermo Scientific Chemicals

Product Code. 11457657
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11457657

Brand: Thermo Scientific Alfa Aesar A15698.14

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-29-1
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00001041
InChI Key IGZGUYVVBABKOY-UHFFFAOYSA-N
Synonym 2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotriflorideShow MoreShow Less
PubChem CID 67957
IUPAC Name 1-iodo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)I

Specifications

Density 1.936
Boiling Point 198°C to 199°C
Flash Point 81°C (177°F)
Refractive Index 1.5305
Quantity 25 g
Beilstein 2090038
Sensitivity Light sensitive
Formula Weight 272.01
Percent Purity 99%
Chemical Name or Material 2-Iodobenzotrifluoride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250416 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15698.14,A15698.22
Lot NumberZ04J059 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog NumberA15698.14,A15698.22
Lot Number10178820 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog NumberA15698.14,A15698.22
Lot NumberS13I044 Certificate TypeCertificate of Analysis Date27/06/2022 Catalog NumberA15698.14,A15698.22
Lot NumberU23H022 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15698.14,A15698.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodobenzotrifluoride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.