Learn More
Zinc-alpha-2-glycoprotein Mouse anti-Human, Clone: 4B8A4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66178-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.Spécification
| Zinc-alpha-2-glycoprotein | |
| Monoclonal | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P25311 | |
| AZGP1 | |
| Zinc-alpha-2-glycoprotein Fusion Protein Ag5661 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4B8A4 | |
| Unconjugated | |
| AZGP1 | |
| AZGP1, ZA2G, ZAG, Zinc alpha 2 glycoprotein, Zinc-alpha-2-glycoprotein, Zn alpha 2 glycoprotein, Zn alpha 2 GP, ZNGP1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 563 | |
| -20°C | |
| Liquid |