Learn More
ZVG™ ZetClean™ Wiper Bowl™ Multitex™ Wipe Dispenser Buckets
ZetClean™ Wiper Bowl™ Multitex™ moist, perforated cleaning wipes are tear-resistant, lint-free, and come in a convenient dispenser bucket. These wipes boast an orange scent.
Marque: ZVG™ 50132-000
Description
The moist Multitex™ wipes from the zetClean™ product range, soaked in a special cleaning solution and highly tear-resistant, are indispensable for cleanliness in workshops and machine parks. They are particularly ideal for workplaces where immediate access to water for cleaning is not available. The soft, tear-resistant viscose wipes with textile-like characteristics are enriched with Vitamin E and aloe vera, making them especially gentle on the skin. They are suitable for removing oils, fats, adhesives, tar, grease, and more.
- Viscose, nonwoven wipes with textile-like characteristics
- Suitable for wet and dry cleaning
- Robust and lint-free
- Tear-resistant
- Soaked in a gentle cleaning solution
- Pleasant fresh orange scent
- Easy tear-off perforation for convenient use
- Resealable dispenser bucket
- 72 wipes per bucket
- Particularly skin-friendly
Spécification
Wiper | |
25 cm | |
25 cm | |
Bleu | |
Non stérile |
9.8 in. | |
9.8 in. | |
Polypropylene, Viscose, Cotton | |
Usage unique |